Learn More
Medchemexpress LLC HY-125877 1mg Medchemexpress, PROTAC Mcl1 degrader-1 CAS:2163793-38-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1258771MG
Encompass_Preferred
Medchemexpress, HY-125877 1mg PROTAC Mcl1 degrader-1 CAS:2163793-38-0 PROTAC Mcl1 degrader-1 (compound C3), a proteolysis targeting chimera (PROTAC), is a potently and selectively Mcl-1 inhibitor with an IC50 of 0.78 μM. PROTAC Mcl1 degrader-1 induces the ubiquitination and proteasomal degradation of Mcl-1 by introducing the E3 ligase cereblon (CRBN)-binding ligand pomalidomide to Mcl-1 inhibitor S1-6 with μM-range affinity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9711