Learn More
New Star Environmental Inc Prote Filter Cartridge (Overflow Protection) for Rocker 300/400/410/430/500
Supplier:  New Star Environmental Inc 16730107
Encompass_Preferred
The Rocker series pumps are equipped with Prote™ overflow protection which can prevent large amounts of liquid from being sucked into the pump. The filter cartridge located in the air inlet removes particles and moisture to prolong the life of the pump.
Catalog No. NC1638257
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.