Learn More
Enzo Life Sciences Proteasome 19S Rpt1/S7 subunit (yeast) polyclonal antibody (25 µl)
Supplier: Enzo Life Sciences BMLPW82550025
Immunogen: Recombinant protein corresponding to the N-terminal 100 amino acids of YTA3. Species reactivity: Yeast. Specificity: Recognizes the Rpt1/S7 subunit of the 19S regulator complex. Applications: WB. Formulation: Liquid. Neat serum.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.