Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Protease Activity Assay Kit, Fluorometric, Green, 500 Tests

Catalog No. 502584938
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-4938 500 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-4938 Supplier Abcam Supplier No. AB112152500TEST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rapidly detect protease activity in cell extracts and biofluids within 1 hr.

  • Assay Type: Enzyme activity
  • Detection Method: Fluorescent
  • Assay Time: 1 hr
  • Sample Types: Cell culture extracts, Other biological fluids, Plasma, Serum
  • Applications: Enzyme activity assay
  • Usage: Suitable for use with a microplate reader
  • Citations: Cited in > 2 publications

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Run Time 1 hr.
Detection Location In-Solution Detection
For Use With (Equipment) Microplate Reader
Format Kit
No. of Reactions 500 Tests
Target Molecule Protease
Detection Method Fluorescence
Sample Type Cell culture extracts, Other biological fluids, Plasma, Serum
Quantity 500 Tests
For Use With (Application) Enzyme activity assay
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.