Learn More
Bioworld Protease Inhibitor Cocktail I for General Use, 1 Vial(s)
Supplier:  Bioworld 220200041
Encompass_Preferred
Lyophilized blend of five inhibitors for inhibiting a broad range of proteases and esterases. Reconstitutes with 1 ml of water to produce a 100X concentrate solution.Specification:Contains: AEBSF, E-64, Leupeptin, Aprotinin, EDTASpecificity: Serine Proteases, Esteroses, Cysteine Proteases, Metalloproteases and Trypsin like ProteasesThis product is sold for laboratory research use only.
Catalog No. 50-252-985
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.