Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Protease (S. aureus)

Catalog No. 89958311 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-311 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-958-311 Supplier Abnova™ Supplier No. P5254
Add to Cart
Add to Cart

Used for Enzyme

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 27kDa
Name Protease (S. Aureus)
Preparation Method Native protein chromatographically purified from S. aureus V8 according to Drapeau, G., Boily, Y., and Houmard, J., J. Biol. Chem., 247, 6720 (1972)
Quantity 1 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name PROTEASE
Assays Protease
Biological Activity ≥500 units per mg dry weight. One Unit causes a change of 0.001 A280 nm per minute at 37°C, pH 7.8 using casein as the substrate.
Species Staphylococcus species
Protein Tag None
Expression System Native protein chromatographically purified from S. aureus V8 according to Drapeau, G., Boily, Y., and Houmard, J., J. Biol. Chem., 247, 6720 (1972)
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.