Learn More
Essendant PROTECSORLAB CS2XLWH30
Supplier:  Essendant ANS44445
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees Offering your team comfortable protective clothing is a must and KleenGuard A40 Liquid and Particle Protection Lab Coats are a great addition Each knee-length lab coat come with serged seams elastic wrists a five-snap closure and no pocket Made of breathable microporous film to keep out debris non-hazardous liquid chemicals and dry particulates the KleenGuard A40 material gives your employees a better liquid barrier than Dupont Tyvek These lab coats pass NFPA 99 for antistatic materials pass ASTM F1670 testing for penetration of blood and bodily fluids and meet the ANSI/ISEA 101-1996 sizing standards Popular industries using this product include aviation pharmaceutical manufacturing liquid handling general manufacturing and so much more Making the right Personal Protection choice is difficult Perform with distinction with
Catalog No. 50-000-22967
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.