Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Protecta™ Comfort Vest-Style Harness

Catalog No. 19070966 Shop All 3M Products
Change view
Click to view available options
Size:
Medium/Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-070-966 Medium/Large
19-070-967 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-070-966 Supplier 3M™ Supplier No. 7012816690
Add to Cart
Add to Cart
This item is not returnable. View return policy

Protecta™ Comfort Vest-Style Harness minimizes workday readjustment and is designed for comfort and convenience

  • Fixed D-ring reduces the need for readjustment throughout the workday
  • Specifically designed for added comfort
  • Moisture-wicking, breathable back padding with soft edging for added comfort
  • Modern design gives harness a fresh and sleek look
  • ANSI-compliant auto-resetting lanyard keepers for fast and easy parking of snap hooks and carabiners
  • Impact indicators make it easy to inspect the harness for prior damaging impact loads
  • Protected labels for long-term identification and inspection
TRUSTED_SUSTAINABILITY

Specifications

Material Polyester
Color Black
Tensile Strength (English) 3600 lb./ANSI
Size Medium/Large
D-Ring Location Back
For Use With (Application) Demolition, Painting, Blasting, Facility Maintenance, Electrical
Product Type Comfort Vest-Style Harness
Webbing Material Polyester
Buckle Style Pass-Thru; Parachute; Tongue Buckle
Capacity (Metric) 190 kg
Capacity (English) 420 lb.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.