Learn More
TerraCycle™ Protection Helmets Zero Waste Box™



Description
- What can you recycle?
- Use this Zero Waste Box to recycle any used Industrial Safety Helmets and Safety Bump Caps. Does not accept other types of protective gear, such as, gloves, hair nets, lab coats, garments, safety goggles, or masks.
- Do not send in medical waste. This includes material contaminated with blood or bodily fluids that originate from health care facilities, hospitals, physicians’ offices, dental offices, blood banks, veterinary clinics, research laboratories, etc.
- How It Works?
- Order: Select a Zero Waste Box and place your order.
- Collect: Collect the accepted items to fill the box.
- Ship: Send it back to TerraCycle with the prepaid return label on your box.
- Reorder: Order your next Zero Waste Box to continue recycling everything.
- What’s Included?
- One Zero Waste Box
- One prepaid return label, affixed to your box
- Processing and recycling of the materials
- Important Shipping Restrictions
- Only send items accepted by this box. Sending items outside the selected category causes contamination and added separation costs.
- Do not send household items considered hazardous, including lithium-ion batteries, pressurized canisters, pesticides, oil-based paint, medical sharps, organics, broken glass, biomedical waste, soiled diapers, or expired medication. Avoid anything sharp, flammable, reactive, corrosive, ignitable, radioactive, toxic, infectious, or pathogenic.
- Do not ship dripping packages. Remove excess product and ensure the box is completely dry before shipping.
Specifications
Specifications
| Capacity (Metric) | 39.66 L |
| Capacity (English) | 10.47 gal. |
| Color | Brown |
| Dimensions (L x W x H) | 11 x 11 x 20 in. |
| Autoclavable | Not Autoclavable |
| Material | Cardboard |
| Product Type | Recycling Box |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.