Learn More
JDM SCIENTIFIC LLC Protective cover for SmartRack
Supplier:  JDM SCIENTIFIC LLC 01609
Encompass_Preferred
The protective hood made of fiber-reinforced polyethylene (mesh film), which was specially designed for the SmartRack , reliably protects the SmartRack against dirt and dust particles as well as providing splash protection against aggressive liquids. The dust and particle protection hood is available in two versions that do not differ in size. As a basic model and as a model with two zippers on the edges of the front. For regular removal of the dust and particle protection hood and for use in unfavorable height conditions, we recommend choosing the model with zippers.
Catalog No. 50-255-8518
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.