Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Poltex Protective Eyewear Holder, 9 Position

Stores protective eyewear in sturdy holder.
$135.70 - $156.90
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Price | Quantity & Availability | |||||
22-000-343
![]() |
Poltex
PROTEYE9M |
Magnet mounted |
Each for $156.90
|
|
|||||
22-000-342
![]() |
Poltex
PROTEYE9W |
Wall mounted |
Each for $135.70
|
|
|||||
Description
Choose from wall mount, or powerful magnet mount. Durable PETG Construction.Specifications
Protective eyewear, 9 positions | |
PETG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title