Learn More
Brady™ Protective Overlaminate for Write-On Labels
Catalog No. 19104532 Shop All Brady Corporation Products
Change view
Click to view available options
Size (English):
6.5 x 4.5 in. (W x H)
7.5 x 5.5 in. (W x H)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (English)
19-104-532 6.5 x 4.5 in. (W x H)
19-104-533 7.5 x 5.5 in. (W x H)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-104-532 Supplier Brady™ Supplier No. 99272
Add to Cart
Add to Cart
Protects handwritten data on vinyl labels from chemicals and abrasion. Brady™ Protective Overlaminate for Write-On Labels help in maintaining a safe work environment.
  • Write-On Arc Flash Labels format
  • Sheets have an acrylic layflat liner
  • Available in 25/Pk. quantity
Specifications
Color Clear
Material Polyester
Product Type Protective Overlaminate for Write-On Label
Size (English) 6.5 x 4.5 in. (W x H)
WARNING: Cancer - www.P65Warnings.ca.gov