Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector XL Laboratory Hood Accessory, Tissue Screen Kit
SDP

Catalog No. 10304179 Shop All Labconco Corporation Products
Change view
Click to view available options
For Use With (Equipment):
Protector XL Hood (36 in.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
10-304-179 Protector XL Hood (36 in.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 10-304-179 Supplier Labconco™ Supplier No. 9576710
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with 3 ft. Protector XL Laboratory Hood. Labconco™ Protector XL Laboratory Hood Accessory, Tissue Screen Kit, helps in trapping tissue wipers and other small, loose materials which may be caught in the airflow.

Screens that mount behind the baffles of chemical fume hood for trapping tissue wipers and other small, loose materials which may be caught in airflow.

Kit components:

  • Mounting hardware
  • Powder-coated stainless steel screen
  • Instructions

Specifications

For Use With (Equipment) Protector XL Hood (36 in.)
Weight (English) 1.8 lb.
Includes Mounting Hardware, Powder-coated Stainless Steel Screen and Instructions
Product Type Tissue Screen Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.