Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein A Antibody Package

Catalog No. 361041369 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-104-1369 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 36-104-1369 Supplier Cytiva Supplier No. 29058805
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in protein purification applications

Protein A package is recommended for the purification of human antibodies. Contents: HiTrap Protein A HP (1 x 1mL) and HiTrap Desalting (1 x 5mL).
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody affinity chromatography
Content And Storage 2°C to 8°C
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.