Learn More
Cytiva Protein A/G HP SpinTrap™ Buffer Kit
Catalog No. 45001522 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
45-001-522 1 Kit
1 options
Catalog No. 45-001-522 Supplier Cytiva Supplier No. 28913567
Add to Cart
Add to Cart
Includes
Sufficient quantities of the following reagents to perform 16 reactions: 10X Binding/Washing buffer, Classic elution buffer, 10X Crosslink elution buffer, 10X Crosslink Solution A, and 10X Crosslink Solution B.
Pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation using Protein A HP, Protein G HP, and Ab SpinTrap™ columns
  • Eliminates time-consuming buffer preparation; allows fast, reproducible enrichment of target protein from complex protein mix
  • Contains reagents for both the Classic and Crosslink protocols (with/without elution of antibody together with target protein, respectively)
  • Convenient format: stock solutions can be diluted directly into the provided bottles for storage
TRUSTED_SUSTAINABILITY
Specifications
Product Type Protein A/G HP SpinTrap Buffer Kit
Description Pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation using Protein A HP, Protein G HP, and Ab SpinTrap columns.
Content And Storage 4°C to 30°C
Quantity 1 Kit