Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein A HP MultiTrap™ Plates

Catalog No. 45001484 Shop All Cytiva Products
Change view
Click to view available options
Sample Volume:
70 to 130 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Volume
45-001-484 70 to 130 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-484 Supplier Cytiva Supplier No. 28903133
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Requires

Four prepacked Protein A HP MultiTrap 96-well filter plates and instructions

For small-scale purification of antibodies or enrichment of proteins of interest by antibody capture in 96-well filter plates

  • Flexible format, with protocols for both classic elution and cross-link methods
  • Formatted for elution for both electrophoresis and LC-MS analysis workflows
  • Fast-binding kinetics and high capacity provide high yield
  • Prepacked with Protein A Sepharose™ High Performance Medium for coupling of antibodies of IgG subclasses
  • Can be used with centrifugation or vacuum, manually or in automated robotic systems
  • Easy scale-up with HiTrap™ Protein A HP prepacked columns

  • Matrix: Highly cross-linked agarose, 6%
  • Binding capacity: approx. 20mg human IgG/mL medium
  • Average particle size: 34μm
  • pH stability: 3 to 9 (long term); 2 to 9 (short term)
  • Well volume: 800μL
  • Centrifugation speed: 700xG
  • Vacuum pressure: -0.5 bar (max.)

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity 20 mg/mL Human IgG Medium
For Use With (Application) Centrifugation, Vacuum
Sample Volume 70 to 130 μL
For Use With (Equipment) For vacuum, centrifugation, manually or with robotic systems
Content And Storage 2°C to 8°C, 20% Ethanol
Quantity 4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.