Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein A HP SpinTrap™ Column

Catalog No. 45001483 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
16 Columns
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-483 16 Columns
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-483 Supplier Cytiva Supplier No. 28903132
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For simple, small-scale antibody purification or enrichment of specific proteins by antibody capture in single-use spin columns

Prepacked with Protein A Sepharose™ High Performance Media for coupling of antibodies of IgG subclasses; used with a standard microcentrifuge

  • Fast binding kinetics and high capacity provide high yield
  • Classic (antigen eluted with antibody) and cross-link (antigen eluted separately) protocols provide flexibility
  • Elution conditions formatted for both electrophoresis and LC-MS analysis workflows
  • Easy scale-up with HiTrap Protein A HP prepacked columns
  • Additional protocol for small-scale purificaiton of monoclonal and polyclonal antibodies from serum and cell culture supernatants

TRUSTED_SUSTAINABILITY

Specifications

Binding Capacity >1 mg Human IgG well
For Use With (Application) Centrifugation
Sample Volume 600 μl
For Use With (Equipment) Standard microcentrifuge.
Content And Storage 2°C to 30°C, 20% Ethanol
Quantity 16 Columns
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.