Learn More
Abcam Protein A, Staphylococcus Aureus, FITC Conjugated, 1 mg
Description
- Protein Type: Native Staphylococcus aureus protein (Protein A)
- Purity: >95%
- Form: Liquid
- Tested Applications: Dot, Flow Cyt, IM, WB
- Purification Technique: Affinity purification Protein A
- Storage Buffer: Preservative: 0.01% Sodium azide; Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Immunoglobulin G |
| Shipping Condition | Blue Ice |
| Substrate Properties | Protein-based Substrate |
| Content And Storage | Store at 4°C. |
| Label or Dye | FITC |
| Reactivity | Staphylococcus aureus |
| Detection Method | Fluorescence |
| Buffer | 1% BSA, 0.88% sodium chloride, 0.424% potassium phosphate, 0.01% sodium azide |
| Concentration | 1 mg/mL |
| Form | Liquid |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.