Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protein APC Recombinant Rabbit Monoclonal Antibody (SC61-03)
GREENER_CHOICE

Catalog No. PIMA541163
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541163 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. PIMA541163 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' MA541163
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Protein APC Recombinant Monoclonal Antibody for IHC (F), IHC (P), Western Blot

This gene encodes a tumor suppressor protein that acts as an antagonist of the Wnt signaling pathway. It is also involved in other processes including cell migration and adhesion, transcriptional activation, and apoptosis. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein APC
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone SC61-03
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Apc
Gene Accession No. P25054, Q61315
Gene Alias adenomatosis polyposis coli; adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli; adenomatous polyposis coli protein; AI047805; APC; APC, WNT signaling pathway regulator; AU020952; AW124434; BTPS2; CC1; Deleted in polyposis 2.5; DP2; DP2.5; DP3; FAP; FPC; GS; mAPC; Min; multiple intestinal neoplasia; PPP1R46; Protein APC; protein phosphatase 1, regulatory subunit 46; RATAPC; truncated adenomatosis polyposis coli; WNT signaling pathway regulator
Gene Symbols Apc
Host Species Rabbit
Immunogen Synthetic peptide within Human Protein APC aa 51-100.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11789, 324
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.