Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protein C Inhibitor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101753 Supplier Invitrogen™ Supplier No. PA5101753
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Serpin A5.

This gene encodes a member of the serine proteinase inhibitor (serpin) superfamily. This member is the principal inhibitor of tissue plasminogen activator (tPA) and urokinase (uPA), and hence is an inhibitor of fibrinolysis. Defects in this gene are the cause of plasminogen activator inhibitor-1 deficiency (PAI-1 deficiency), and high concentrations of the gene product are associated with thrombophilia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein C Inhibitor
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SERPINA5
Gene Accession No. P05154, P70458
Gene Alias 4933415L04; acrosomal serine protease inhibitor; alpha-1 antiproteinase; antitr; antitrypsin; LOC480424; LOC483954; PAI3; PAI-3; Pci; PCI-B; PLANH3; Plasma serine protease inhibitor; plasminogen activator inhibitor 3; plasminogen activator inhibitor III; plasminogen activator inhibitor-3; PROCI; Protein C inhibitor; serine (or cysteine) peptidase inhibitor, clade A, member 5; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serine (or cysteine) proteinase inhibitor, clade A, member 5; serpin A5; serpin B3; serpin family A member 5; serpin family B member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 5; serpin peptidase inhibitor, clade A, member 5; serpin peptidase inhibitor, clade B (ovalbumin), member 4; SERPINA5; SERPINB3; SERPINB4
Gene Symbols SERPINA5
Host Species Rabbit
Immunogen A synthesized peptide derived from human SERPINA5(Accession P05154), corresponding to amino acid residues G310-T360.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 268591, 5104, 65051
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.