Learn More
Siemens Healthineers Protein C Reagent
Catalog No. 23044681 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
4 x 1 mL/Kit
Catalog No. Quantity
23-044-681 4 x 1 mL/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-044-681 Supplier Siemens Healthineers Supplier No. 10446185
Add to Cart
Add to Cart
This item is not returnable. View return policy
Coagulation test for the quantitative determination of protein C activity in human plasma.
Protein C is a coagulation test used for the quantitative determination of protein C activity in human plasma. The reagent is suitable for the detection of hereditary or acquired protein C deficiencies.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Thrombophilia Testing
Quantity 4 x 1 mL/Kit