Learn More
Abcam Protein Carbonyl ELISA Kit, 32 tests
Supplier:  Abcam AB23853632TEST
Encompass_Preferred
Assay type: Cell-based (quantitative)Detection method: ColorimetricPlatform: Microplate readerSample type: Cell Lysate, EDTA Plasma, Hep Plasma, Serum, Tissue Lysate
Catalog No. NC2610149
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.