Learn More
Abcam Assay type: Cell-based (quantitative)Detection method: ColorimetricPlatform: Microplate readerSample type: Cell Lysate, EDTA Plasma, Hep Plasma, Serum, Tissue Lysate
Supplier:  Abcam AB23853696TEST
Encompass_Preferred
Assay type: Cell-based (quantitative)Detection method: ColorimetricPlatform: Microplate readerSample type: Cell Lysate, EDTA Plasma, Hep Plasma, Serum, Tissue Lysate
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2610151