Learn More
Viva Products BJP-5/40 Clinical Concentrator, with pipettes
SDP
Supplier:  Viva Products BJP5/40P
Encompass_Preferred
Clinical Ultrafiltration Concentrator; For concentration of proteins in clinical diagnostic samples such as urine and spinal fluid; Used prior to electrophoresis of monoclonal immunoglobulins (M-proteins) as well as immunofixation electrophoresis (IFE); Sample volume to 5 ml; Final concentrate volume as low as 50 microliters for 100X concentration factor; 7,500 MWCO; Includes disposable pipettes; Number of tests is 40/PK
Catalog No. 50-355-736
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.