Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Protein G, Alexa Fluor™ 488 conjugate

Catalog No. P11065
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P11065 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P11065 Supplier Invitrogen™ Supplier No. P11065
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protein G, Alexa Fluor™ 488 conjugate

Alexa Fluor 488 protein G conjugate exhibits bright, green fluorescence (absorption/emission ∼495/519 nm). Protein G is a bacterial protein similar to protein A but with superior selectivity for mouse and human IgG.

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Alexa Fluor 488
Quantity 1 mg
Product Line Alexa Fluor
Form Lyophilized Powder
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.