Learn More
Invitrogen™ protein G, horseradish peroxidase conjugate
Description
The horseradish peroxidase (HRP) conjugate of protein G can be used to detect the Fc portion of various classes and subclasses of immunoglobulins from a variety of species. Protein G is a bacterial protein similar to protein A but with superior selectivity for mouse and human IgG.
Specifications
Specifications
| Conjugate | HRP |
| Quantity | 1 mg |
| Form | Lyophilized Powder |
| Content And Storage | Store in freezer (-5 to -30°C). |
| Shipping Condition | Room Temperature |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.