Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein G HP MultiTrap™ Plates

Catalog No. 45001486 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
4 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-486 4 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-001-486 Supplier Cytiva Supplier No. 28903135
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Four prepacked Protein G HP MultiTrap 96-well filter plates with instructions, plus a protocol, with no pretreatment of sample required, for efficient, small-scale purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants in less than 20 min.

For small-scale purification of antibodies or enrichment of proteins of interest in prepacked 96-well filter plates

  • Flexible format, with protocols for both classic elution and cross-link methods
  • Formatted for elution for both electrophoresis and LC-MS analysis workflows
  • Fast-binding kinetics and high capacity provide high yield
  • Prepacked with Protein G Sepharose™ High Performance Medium for coupling of antibodies of IgG subclasses
  • Can be used with centrifugation or vacuum, manually or in automated robotic systems
  • Easy scale-up with HiTrap™ Protein G HP prepacked columns

  • Matrix: Highly cross-linked agarose, 6%
  • Binding capacity: approx. 25mg human IgG/mL medium (0.5 to 1mg IgG/well)
  • Average particle size: 34μm
  • pH stability: 3 to 9 (long term); 2 to 9 (short term)
  • Well volume: 800μL
  • Centrifugation force: <700 x G
  • Vacuum pressure: -0.5 bar (max.)

TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein G Sepharose HP MultiTrap plate
Quantity 4 Pack
Includes 4 Prepacked Protein G HP MultiTrap 96-well filter plates with instructions, plus a proto col
Material Polypropylene, Polyethylene
Ligand Type Recombinant Protein G lacking Albumin-binding region (2mg/mL medium)
Max. Pressure -0.5 bar
pH Range 3 to 9 (long term), 2 to 9 (short term)
Particle Size 34μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 2° to 8°C
Column Type Protein G Sepharose Plate
For Use With (Application) Small-scale purification of Antibodies or enrichment of Proteins of interest in prepacked 96-well filter plates that are ready to use, electrophoresis or LC/MS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.