Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva Protein G HP SpinTrap™ Columns
GSA_VA

Catalog No. 45001485 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
800 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-001-485 800 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 45-001-485 Supplier Cytiva Supplier No. 28903134
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

16 prepacked Protein G HP SpinTrap columns, one plastic bottom cap removal tool, and instructions. Also included is a protocol, with no pretreatment of sample required, for efficient, small-scale purification of monoclonal and polyclonal antibodies from serum and cell culture supernatants in less than 20 min.

For small-scale purification of antibodies or enrichment of proteins of interest in prepacked spin columns

  • Protocols for both classic elution and cross-link methods; designed for use with a standard microcentrifuge
  • Formatted for elution for both electrophoresis and LC-MS analysis workflows
  • Fast-binding kinetics and high capacity provide high yield
  • Prepacked with Protein G Sepharose™ High Performance Medium for coupling of antibodies of IgG subclasses
  • Easy scale-up with HiTrap™ Protein G HP prepacked columns

  • Matrix: Highly cross-linked agarose, 6%
  • Binding capacity: >1mg human IgG/column
  • Average particle size: 34μm
  • pH stability: 3 to 9 (long term); 2 to 9 (short term)
  • Column volume: 800μL
  • Max. sample loading volume: 600μL

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein G Sepharose HP SpinTrap spin column
Quantity 800 μL
Includes 16 Prepacked Protein G HP SpinTrap column, one Plastic botto m cap removal to ol and instructions, proto col
Ligand Type Recombinant Protein G lacking Albumin-binding region (2mg/mL medium)
pH Range 3 to 9 (long term), 2 to 9 (short term)
Particle Size 34μm
Support Type 6% Highly Cross-linked Agarose
Content And Storage 4° to 8°C
Column Type Protein G Sepharose Spin Column
For Use With (Application) Small-scale purification of Antibodies or enrichment of Proteins of interest in prepacked spin column, electrophoresis or LC/MS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.