Learn More
Genscript Corporation Protein G MagBeads
Supplier:  Genscript Corporation L00274
Encompass_Preferred
GenScript Protein G MagBeads are superparamagnetic beads covalently bound with recombinant protein G for antibody purification. The beads are supplied as 25% slurry in phosphate buffered saline (PBS), pH 7.4, containing 20% ethanol. The Protein G MagBeads have a binding capacity of more than 10 mg Goat IgG per 1 ml settled beads.
Catalog No. NC2101161
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.