Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein G Polyclonal Antibody, Agarose, Rockland™
SDP

Catalog No. 501991064
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mg
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1064 25 μL
50-199-1063 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-1064 Supplier Rockland Immunochemicals Supplier No. 2004191S
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at 4°C prior to opening. DO NOT FREEZE. Anti-Protein G antibody is an IgG fraction antibody coupled to activated agarose. Sufficient antibody capacity is provided to bind a minimum of 5 mg of Protein G.

Specifications

Antigen Protein G
Applications Western Blot
Classification Polyclonal
Concentration 10 mg/mL
Conjugate Agarose
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Host Species Rabbit
Immunogen Protein G (Streptococcus sp.)
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Slurry
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.