Learn More
Sartorius Octet Protein G (ProG) Biosensors, Rapid Quantification of Mammalian IgGs, Pre-Immobilized Protein G, Case (20 trays)
Catalog No. NC1970229 Shop All Sartorius Products
Encompass
The Octet Protein G (ProG) Biosensors provide rapid and direct quantification of mammalian IgGs, enhancing bioprocess efficiency. With pre-immobilized Protein G, they outperform Protein A, making them ideal for applications like hybridoma screening. Their regenerable format ensures cost-effectiveness and minimal sample handling, delivering precise results quickly. Compatible with Octet systems, they facilitate seamless antibody concentration determination, offering a dynamic range from 0.05 to 2000 µg/mL.
Catalog No. NC1970229 Supplier Sartorius Supplier No. 185084
Add to Cart
Add to Cart
Request a Quote
The Octet Protein G (ProG) Biosensors provide rapid and direct quantification of mammalian IgGs, enhancing bioprocess efficiency. With pre-immobilized Protein G, they outperform Protein A, making them ideal for applications like hybridoma screening. Their regenerable format ensures cost-effectiveness and minimal sample handling, delivering precise results quickly. Compatible with Octet systems, they facilitate seamless antibody concentration determination, offering a dynamic range from 0.05 to 2000 µg/mL.