Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Protein Gene Product 9.5 (PGP9.5) Ab-1, Mouse Monoclonal Antibody

Catalog No. ms1760r1 Shop All Epredia Products
Change view
Click to view available options
Quantity:
1 mL
Form:
Prediluted
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Form
MS1760R1 1 mL Prediluted
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MS1760R1 Supplier Epredia™ Supplier No. MS1760R1

Provide accurate, reproducible results with the Epredia™ Protein Gene Product 9.5 (PGP9.5) Ab-1, Mouse Monoclonal Antibody.

Protein gene product 9.5 is a ubiquitin C-terminal hydroxylase found in neurons and nerve fibres of central and peripheral nervous system; many neuroendocrinal cells, segments of renal tubules, spermatogonia, leydig cells of testis; and ova. It is present in cellular inclusions and can be a useful as a neuronal marker and in the studies of neurodegenerative disorders.

Host Species: Mouse

Clone: 10A1

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein corresponding to full-length human protein gene product 9.5 molecule

Molecular Weight: 25-27kDa

Positive Control: Small intestine

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

General References:

1. Wilkinson K D, et al. (1989) Science, 246:670-675.

2. Wilson P O G, et al. (1988) Br J Exp Pathol, 69:91-104.

Specifications

Antigen Protein Gene Product 9.5 (PGP9.5) Ab-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 10A1
Conjugate Unconjugated
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human protein gene product 95 molecule
Quantity 1 mL
Regulatory Status IVD
Research Discipline Neuroscience
Primary or Secondary Primary
Target Species Human
Form Prediluted
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.