Learn More
Cell Biolabs Inc Copper Oxidized HumanLow Density Lipoprotein (LDL), 100 µg
Supplier:  Cell Biolabs Inc STA214
Encompass_Preferred
Cell Biolabs' Copper (Cu++) Oxidized Human Low Density Lipoprotein (LDL) is provided as 100 ug at 1 mg/mL in PBS, pH 7.5, containing 0.5 mM EDTA and 0.02% sodium azide.
Catalog No. 50-673-472
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.