Learn More
ProteinSimple Protein Normalization Module
Supplier:  ProteinSimple DMPN02
Encompass
The Protein Normalization Module from Simple Western is an accessory that enables automated normalization of protein levels across multiple samples for more accurate and reproducible western blot data. It works by probing a housekeeping target like actin or GAPDH in parallel with the protein of interest. The module consists of two chambers that allow simultaneous probing of the normalization protein and target protein. It is compatible with Wes, Simple Western's capillary electrophoresis and blotting system. The module delivers more accurate normalization than typical strip-based methods.
Catalog No. NC2075413