Learn More
Siemens Healthineers Protein S Ac Test Kit
Catalog No. 23044666 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
2 x 2 mL/Kit
Catalog No. Quantity
23-044-666 2 x 2 mL/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-044-666 Supplier Siemens Healthineers Supplier No. 10445968
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determination of the functional activity of protein S in human plasma.
Protein S Ac, a coagulometric activity reagent, is used for the detection of hereditary or acquired protein S deficiencies.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Thrombophilia Testing
Quantity 2 x 2 mL/Kit