Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PROTEIN S Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590849
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590849 Supplier Invitrogen™ Supplier No. PA590849
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDINKPGPLF KPENGLLETK VYFAGFPRKV ESELIKPINP RLDGCIRSWN LMKQGASGIK EIIQEKQNKH CLVTVEKGSY YPGSGIAQFH IDYNNVSSAE GWHVNVTLNI RPSTGTGVML ALVSGNNTVP FAVSLVDSTS EKSQDILLSV ENTVIYRIQA LSLCSDQQSH LEFRVNRNNL ELSTPLKIET ISHEDLQRQL AVLDKAMKAK VATYLGGLPD VPFSATPVNA FYNGCMEVNI NGVQLDLDEA ISKHNDIRAH SCPSVWKKTK NS; Positive Samples: LO2, HeLa, 293T, Mouse liver, Mouse kidney, Rat liver, Rat kidney.

Protein S is a vitamin K-dependent, potent neutral anticoagulant plasma glycoprotein. Protein S has a molecular weight of approximately 70 kDa and consists of a single polypeptide chain. The concentration of protein S in human plasma is 25 μg/mL, with a half-life of approximately two days. About 40% of Protein S in plasma is in a free form, whereas 60% is complexed with C4b-binding Protein (C4BP). Only the free Protein S functions as a cofactor to antigen presenting cells (APC) in the APC-dependent degradation of Factor Va and Factor VIIIa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PROTEIN S
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.74 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PROS1
Gene Accession No. P07225, P53813, Q08761
Gene Alias AW214361; Preproprotein S; PROS; PROS 1; PROS1; protein S; protein S (alpha); Protein S alpha; protein Sa; proteinS; PS 21; PS 22; PS 23; PS 24; PS 25; PS 26; PS21; PS22; PS23; PS24; PS25; PS26; PSA; rabbit protein S; THPH5; THPH6; unnamed protein product; vitamin K-dependent plasma protein S; vitamin K-dependent protein S; vitamin K-dependent protein S precursor
Gene Symbols PROS1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 405-676 of human PROS1 (NP_0003042).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19128, 5627, 81750
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.