Learn More
Cytiva™ Protein Select™ Resin
Catalog No. 09940102 Shop All Cytiva Products
Change view
Click to view available options
Volume (Metric):
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
09-940-102 25 mL
09-940-103 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-940-102 Supplier Cytiva™ Supplier No. 17542101
Only null left
Add to Cart
Add to Cart
An affinity chromatography resin for purifying recombinant proteins using the self-cleaving Cytiva™ Protein Select™ tag.
  • Simple affinity purification protocol for recombinant proteins
  • One-step purification and tag cleavage without the need for proteases or other elution reagents
  • Delivers a highly pure, native protein with no remaining tag amino acids after tag cleavage
  • Can be processed under mild conditions, which means no exposure to conditions that cause protein aggregation or loss of biological activity
  • Flexible, enabling the use in multiple expression systems
TRUSTED_SUSTAINABILITY
Specifications
Type Resin
Quantity 1 Ea.
Form Resin Slurry
Column Type Affinity Column
Volume (Metric) 25 mL