Learn More
Abcam Protein Synthesis Assay Kit, Red, 100/Pk.
Description
- Enables labeling of cells undergoing translation with red fluorescent dye
- Utilizes the O-Propargyl-Puromycin method
- Detection method: Fluorescent
- Suitable sample types: Suspension cells, Adherent cells
- Compatible with flow cytometer or fluorescence microscope
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Protein |
| No. of Reactions | 100 Tests |
| Label or Dye | Red Fluorescent Dye |
| Detection Location | In-Cell Detection |
| For Use With (Equipment) | Flow Cytometer, Fluorescence Microscope |
| Format | Kit |
| Detection Method | Fluorescence |
| Sample Type | Suspension cells, Adherent cells |
| Quantity | 100 Tests |
| For Use With (Application) | Flow Cytometry, Fluorescence Microscopy |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.