Learn More
Novus Biologicals™ Protein Tyrosine Phosphatase Activity Assay Kit (Fluorometric)

Description
Detection method: Fluorescence (Ex/Em = 368/440 nm). Applications: Rapid assessment of PTPase activity in biological samples or recombinant PTPase preparations.
Kit Contents:
- PTPase Assay Buffer
- PTPase Inhibitor (Suramin)
- PTPase Positive Control
- PTPase Substrate
- Fluorescent Standard
- Disulfide Reducing Agent (DTT)
Specifications
Specifications
| Quantity | 100 Assays |
| Product Type | Activity Assay Kit (Fluorometric) |
| Content And Storage | Store at -20°C in the dark. Avoid freeze-thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.