Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Protein Z Monoclonal Antibody (OTI4F1)
GREENER_CHOICE

Catalog No. PIMA526806
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526806 Supplier Thermo Scientific Supplier No. MA526806
Only null left
Add to Cart
Add to Cart

Protein Z Monoclonal Antibody (OTI4F1)

Protein Z Monoclonal Antibody for Western Blot, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen Protein Z
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4F1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PROZ
Gene Accession No. P22891
Gene Alias protein Z, vitamin K dependent plasma glycoprotein; protein Z, vitamin K-dependent plasma glycoprotein; PROZ; PZ; vitamin K-dependent protein Z; vitamin K-dependent protein Z precursor variant 1
Gene Symbols PROZ
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 115-400 of PROZ produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8858
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.