Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Proteinase K
SDP

Catalog No. NB90066727 Shop All Novus Products
Change view
Click to view available options
For Use With (Application):
Preparation of Working Solutions 1. Place 100 uL Proteinase K Solution in a Glass Tube, Add 2.4 mL Distilled Water, and Mix. Protocol Recommendations 1. Optimal Digestion Conditions Will Vary Depending on Fixation Conditions and Should be Optimized by The End User. 2. Incubation of Tissue Sections for 10-30 Minutes (at 37 C is Sufficient for in Situ.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
NB90066727 Preparation of Working Solutions 1. Place 100 uL Proteinase K Solution in a Glass Tube, Add 2.4 mL Distilled Water, and Mix. Protocol Recommendations 1. Optimal Digestion Conditions Will Vary Depending on Fixation Conditions and Should be Optimized by The End User. 2. Incubation of Tissue Sections for 10-30 Minutes (at 37 C is Sufficient for in Situ.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB90066727 Supplier R&D Systems™ Supplier No. NB90066727
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems.

This is a 25X concentrated solution. For use, prepare a working solution: Place 100 uL Proteinase K solution in a glas tube, add 2.4 mL distilled water and mix. The optimal digestion conditions will vary depending on the fixation conditions and should be optimized by individual laboratory. However, incubation of tissue sections for 10-30 minutes at 37C is sufficient for in situ.

Specifications

Formulation Sodium Azide is included at less than 0.1%. Exact amount not available.
Product Line Novus
For Use With (Application) Preparation of Working Solutions 1. Place 100 uL Proteinase K Solution in a Glass Tube, Add 2.4 mL Distilled Water, and Mix. Protocol Recommendations 1. Optimal Digestion Conditions Will Vary Depending on Fixation Conditions and Should be Optimized by The End User. 2. Incubation of Tissue Sections for 10-30 Minutes (at 37 C is Sufficient for in Situ.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.