Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Proteinase K from Tritirachium album

Catalog No. ICN19398110 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
100 mg
500 mg
1 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19398110 10 mg
ICN19398105 5 mg
ICN19398125 25 mg
ICN19398180 100 mg
ICN19398190 500 mg
ICN19398191 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. ICN19398110 Supplier MP Biomedicals™ Supplier No. 0219398110
Add to Cart
Add to Cart

Highly active stable endopeptidase with a broad spectrum of action

Proteinase K is used in digestion of proteins from brain tissue samples for prions in Transmissible Spongiform Encephalopathies (TSE) research. Useful for the proteolytic inactivation of nucleases during the isolation of DNA and RNA, as well as in removal of endotoxins bound to cationic proteins such as lysozyme and ribonuclease A and in determination of enzyme localization on membranes.

  • Grade: Molecular biology reagent
  • Specific Activity: >30 mAnson u/mg
  • Presentation: White powder
  • Isoelectric Point: 8.9 (Lit.)
  • Extinction Coefficient: E1%= 14.2 (280 nm,10 mM NaCl and 5 mM CaCl2, pH 8.0) (Lit.)
  • pH: 7.5 -12.00 (denatured hemoglobin as substrate) (Lit.)
  • Soluble in water

One Anson unit is the amount of enzyme which will liberate 1 °mole of folin-positive amino acids per minnute at pH 7.5 and 35°C using hemoglobin as substrat

Isolation of DNA and RNA

TRUSTED_SUSTAINABILITY

Specifications

Color White
Grade Molecular Biology
pH 7.5 to 12
For Use With (Application) Useful for the proteolytic inactivation of Nucleases during the isolation of DNA and RNA
Content And Storage 4°C
Source Tritirachium album
Quantity 10 mg
Product Type Proteinase K
Molecular Weight (g/mol) 30000
Inhibitors Diisopropylfluorophosphate or Phenylmethane Sulfonyl Fluoride, AEBSF and Trypsin
Form Lyophilized Powder
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.