Learn More
Gold Biotechnology Inc PROTEINASE K SOL 20MG
SDP
Supplier:  Gold Biotechnology Inc P480SL10
Encompass
Proteinase K is a highly reactive nonspecific serine protease that belongs to the subtilisin family of proteins. It cleaves at the carboxylic acid side of aliphatic, aromatic, or hydrophobic amino acids. Proteinase K is capable of inactivating RNases and DNases and is used in the isolation or preparation of high molecular weight nucleic acids. Proteinase K is also useful for helping to characterize enzymes, due to its cleavage specificity. This enzyme was designated proteinase K because of its ability to hydrolyze keratin. Proteinase K is stable in a wide variety of detergents and buffer salts and at various temperatures and pH. The isoelectric point of proteinase K is 8.9.
Catalog No. NC1956504
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.