Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Proteinase K Solution, 600mAU/mL

Catalog No. 710493 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-049-3 2 mL
71-049-4 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-049-3 Supplier MilliporeSigma™ Supplier No. 710493
Add to Cart
Add to Cart

Efficient removal of proteins from nucleic acid solutions

  • Proteinase K is a highly active 28,904-Da serine protease isolated from the fungus Tritirachium album
  • The enzyme exhibits broad cleavage specificity on native and denatured proteins and is widely used in the purification of DNA and RNA
  • Its activity is increased in the presence of denaturants such as SDS (1%) and elevated temperature (50-60°C)
  • The recommended working concentration is 50-100μg/mL for protein removal and enzyme inactivation, and up to 2mg/mL for tissue treatment
  • The Proteinase K, Lyophilized powder can be prepared as a 20mg/mL stock solution in water and stored in aliquots at -20°C
  • The enzyme is also available as a ready-to-use concentrated stock solution (600mAU/mL) that is convenient for routine use in most applications
  • 1 mg of Proteinase K is the equivalent of 30mAU (AU = Anson unit)
  • The Novagen Proteinase K products are free of detectable DNase and RNase
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Quantity 2 mL
Product Type Proteinase K
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.