Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Proteinase K (T. album)

Catalog No. 89958318 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-958-318 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-958-318 Supplier Abnova™ Supplier No. P5281
Add to Cart
Add to Cart

Used for Enzyme

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 28.9kDa
Name Proteinase K (T. album)
Preparation Method Native protein purified from Tritirachium album limber
Quantity 25 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name PROTEINASE
Assays Isolation of intact DNA or RNA
Biological Activity ≥20 units per mg dry weight. One Unit releases one micromole of Folin positive amino acids, measured as tyrosine, at 37°C, pH 7.5, using urea denatured hemoglobin as the substrate.
Species Fungi
Protein Tag None
Expression System Native protein purified from Tritirachium album limber
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.