Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Proteinase K, Tritirachium album

Catalog No. 539480100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25mg
100mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-948-0100MG 100mg
53-948-025MG 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 53-948-0100MG Supplier MilliporeSigma™ Supplier No. 539480100MG
Add to Cart
Add to Cart

Serine protease that exhibits strong proteolytic activity on a wide variety of denatured and native proteins of high molecular weight

  • Because of its rapid proteolytic inactivation of endogenous nucleases, it can be used for isolation of mRNA and high molecular weight DNA
  • Preferentially cleaves bonds next to the carbonyl group of N-substituted hydrophobic, aliphatic, and aromatic amino acids
  • Inhibited by DFP, Hg2+, and PMSF
  • Not inactivated by metal chelators, sulfhydryl reagents, TLCK, or TPCK
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from calcium acetate
Quantity 100mg
Format Lyophilized solid
pH Optimum 7.5 - 12.0
Content And Storage Store at+2°C to+8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.