Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ ProteoExtract™ Native Cytoskeleton Enrichment

Catalog No. 1710210 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
32 Assays/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
17-102-10 32 Assays/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-102-10 Supplier MilliporeSigma™ Supplier No. 1710210
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • One bottle containing 1.9mL of 10x Cellular Extraction Buffer
  • One bottle containing 3.5mL of 20x Cytoskeleton Wash Buffer
  • One bottle containing 9mL of ready to use Nuclear Extraction Buffer
  • One vial containing 100μL of 1000x Protease Inhibitor Cocktail solution
  • One vial containing 150μL of 1000x Sodium Orthovanadate solution
  • One vial containing 50μL of 200x donkey anti-mouse FITC conjugate
  • One bottle containing 12.5mL of 10x Blocking/Permeabilization Buffer
  • One vial containing 15μg lyophilized TRITC-Conjugated Phalloidin
  • One vial containing 100μL of 200x DAPI
  • One vial containing 100μL of 100x anti-Vinculin mouse monoclonal IgG1 antibody
  • One vial containing 16μL of 500x anti-GAPDH mouse monoclonal IgG1 antibody

Requires

  • Pipettors and tips capable of accurately measuring 1-1000μL
  • Graduated serological pipettes
  • Distilled or deionized water
  • 4% paraformaldehyde solution for fixing cells
  • 1x Dulbecco's Phosphate Buffered Saline (D-PBS)
  • Cover slips
  • Mounting fluid

This method provides means to directly study cytoskeleton-associated pool of proteins in normal and diseased cytoskeletons

  • Provides cytoskeleton purification detergent buffers, sufficient for extraction from ten 100mm culture dishes to retain focal adhesion and actin-associated proteins while removing soluble cytoplasmic and nuclear proteins from the cell.
  • Anti-Vimentin and anti-GAPDH antibodies are provided as markers for cytoskeleton and cytosol, respectively, in western blot analysis
  • This kit will greatly increase the ability to detect and study the low abundance actin-associated proteins which are typically masked in conventional methods of Western blotting analysis of whole cell lysates
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage The kit may be used for up to 4 months after receipt. Avoid repeated freeze/thaw cycles, and aliquot if necessary
Detection Method Fluorescence
For Use With (Application) Immunofluorescence, Immunocytochemistry
Quantity 32 Assays/Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.