Learn More
R&D Systems™ Proteome Profiler Human Angiogenesis Array Kit
GREENER_CHOICESDP
Catalog No. ARY007 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY007 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY007 Supplier R&D Systems™ Supplier No. ARY007
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Proteome Profiler Human Angiogenesis Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Angiogenesis Array Kit contains 4 membranes - each spotted in duplicate with 55 different angiogenesis antibodies.
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit