Learn More
R&D Systems™ Proteome Profiler Human Pluripotent Stem Cell Array Kit
GREENER_CHOICESDP
Catalog No. ARY010 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY010 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY010 Supplier R&D Systems™ Supplier No. ARY010
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
The Proteome Profiler Human Pluripotent Stem Cell Array Kit requires no specialized equipment and eliminates the need for multiple Western blot experiments.
The Proteome Profiler Human Pluripotent Stem Cell Array Kit contains 8 membranes - each spotted in duplicate with 15 different stem cell marker antibodies.
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit