Learn More
R&D Systems™ Proteome Profiler Human Protease Inhibitor Array Kit
GREENER_CHOICESDP
Catalog No. ARY023 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
ARY023 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. ARY023 Supplier R&D Systems™ Supplier No. ARY023
Customers require a valid business account associated to their profile to access pricing and ordering options.
This item is not returnable. View return policy
Rapid, sensitiv, and economical tool to simultaneously detect the relative changes of 32 protease inhibiting analytes in a single sample
The Proteome Profiler Human Protease Inhibitor Array Kit contains 4 membranes - each spotted in duplicate with 32 different protease inhibitor antibodies.
Specifications
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Line R&D Systems
Quantity 1 Kit